missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chlorobenzoate, 98%

Catalog No. p-7080549
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1014114 25 g
AAA1014122 100 g
AAA1014136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1014114 Supplier Thermo Scientific Chemicals Supplier No. A1014114

CAS: 7335-25-3 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7335-25-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00016338
InChI Key RETLCWPMLJPOTP-UHFFFAOYSA-N
Synonym 2-chlorobenzoic acid ethyl ester, ethyl o-chlorobenzoate, benzoic acid, 2-chloro-, ethyl ester, ethyl-2-chlorobenzoate, ethyl2-chlorobenzoate, pubchem3703, acmc-1bfxj, ethyl 2-chloranylbenzoate, chlorobenzoic acid ethyl ester, ksc498a5r
PubChem CID 81783
IUPAC Name ethyl 2-chlorobenzoate
SMILES CCOC(=O)C1=CC=CC=C1Cl

Specifications

Density 1.194
Boiling Point 243°C
Flash Point >110°C (230°F)
Refractive Index 1.524
Quantity 25 g
Beilstein 2207359
Formula Weight 184.62
Percent Purity 98%
Chemical Name or Material Ethyl 2-chlorobenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-842-6
TSCA Yes

RUO – Research Use Only