missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Chloroacetoacetate 95.0+%, TCI America™
$42.36 - $309.95
Chemical Identifiers
| CAS | 609-15-4 |
|---|---|
| Molecular Formula | C6H9ClO3 |
| Molecular Weight (g/mol) | 164.59 |
| MDL Number | MFCD00009141 |
| InChI Key | RDULEYWUGKOCMR-UHFFFAOYNA-N |
| PubChem CID | 11858 |
| IUPAC Name | ethyl 2-chloro-3-oxobutanoate |
| SMILES | CCOC(=O)C(Cl)C(C)=O |
Chemical Identifiers
| 609-15-4 | |
| 164.59 | |
| RDULEYWUGKOCMR-UHFFFAOYNA-N | |
| ethyl 2-chloro-3-oxobutanoate |
| C6H9ClO3 | |
| MFCD00009141 | |
| 11858 | |
| CCOC(=O)C(Cl)C(C)=O |
Specifications
| 609-15-4 | |
| 77°C | |
| MFCD00009141 | |
| 2920 | |
| RDULEYWUGKOCMR-UHFFFAOYNA-N | |
| ethyl 2-chloro-3-oxobutanoate | |
| 11858 | |
| ≥95.0% (GC) | |
| Ethyl 2-Chloroacetoacetate |
| Yellow | |
| C6H9ClO3 | |
| 25 g | |
| ethyl 2-chloroacetoacetate, 2-chloroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-3-oxo-, ethyl ester, ethyl alpha-chloroacetoacetate, ethyl-2-chloracetoacetate, 2-chloro-3-oxobutyric acid ethyl ester, ethyl2-chloroacetoacetate, 2-chloro-3-oxobutanoic acid, ethyl ester, ethyl2-chloro-3-oxobutanoate, ethyl 2-chloro-3-oxobutyrate | |
| CCOC(=O)C(Cl)C(C)=O | |
| 164.59 | |
| 164.59 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2760
TSCA : Yes