accessibility menu, dialog, popup

UserName

H3COOClCH3O

CAS RN 609-15-4

IUPAC Name: ethyl 2-chloro-3-oxobutanoate
Synonyms: ethyl 2-chloro-3-oxobutanoate
Molecular Weight (g/mol): 164.59
Molecular Formula: C6H9ClO3
InChi Key: RDULEYWUGKOCMR-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Cl)C(C)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)