missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Chloroacetoacetate 95.0+%, TCI America™

Catalog No. C1218500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1218500G 500 g
C121825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1218500G Supplier TCI America Supplier No. C1218500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 609-15-4
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00009141
InChI Key RDULEYWUGKOCMR-UHFFFAOYNA-N
PubChem CID 11858
IUPAC Name ethyl 2-chloro-3-oxobutanoate
SMILES CCOC(=O)C(Cl)C(C)=O

Specifications

Color Yellow
Boiling Point 77°C
Quantity 500 g
UN Number 2920
Synonym ethyl 2-chloroacetoacetate, 2-chloroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-3-oxo-, ethyl ester, ethyl alpha-chloroacetoacetate, ethyl-2-chloracetoacetate, 2-chloro-3-oxobutyric acid ethyl ester, ethyl2-chloroacetoacetate, 2-chloro-3-oxobutanoic acid, ethyl ester, ethyl2-chloro-3-oxobutanoate, ethyl 2-chloro-3-oxobutyrate
Formula Weight 164.59
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Chloroacetoacetate
EINECSNumber (2)-2760
TSCA Yes