missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Chloroacetoacetate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 77°C |
| Quantity | 25 g |
| UN Number | 2920 |
| Synonym | ethyl 2-chloroacetoacetate, 2-chloroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-3-oxo-, ethyl ester, ethyl alpha-chloroacetoacetate, ethyl-2-chloracetoacetate, 2-chloro-3-oxobutyric acid ethyl ester, ethyl2-chloroacetoacetate, 2-chloro-3-oxobutanoic acid, ethyl ester, ethyl2-chloro-3-oxobutanoate, ethyl 2-chloro-3-oxobutyrate |
| Formula Weight | 164.59 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl 2-Chloroacetoacetate |
Safety and Handling
| EINECSNumber | (2)-2760 |
| TSCA | Yes |