Learn More
Ethyl 2-chloroacetoacetate, 96%
CAS: 609-15-4 | C6H9ClO3 | 164.59 g/mol
Supplier: Thermo Scientific Chemicals A1981036
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ethyl 2-chloroacetoacetate | |
| <-80°C | |
| 200°C | |
| 0.96 | |
| 1.4425 | |
| 500 g | |
| ethyl 2-chloroacetoacetate, 2-chloroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-3-oxo-, ethyl ester, ethyl alpha-chloroacetoacetate, ethyl-2-chloracetoacetate, 2-chloro-3-oxobutyric acid ethyl ester, ethyl2-chloroacetoacetate, 2-chloro-3-oxobutanoic acid, ethyl ester, ethyl2-chloro-3-oxobutanoate, ethyl 2-chloro-3-oxobutyrate | |
| CCOC(=O)C(Cl)C(C)=O | |
| 164.59 | |
| 164.59 | |
| Liquid |
| 609-15-4 | |
| 1.182 | |
| 54°C (180°F) | |
| C6H9ClO3 | |
| MFCD00009141 | |
| 774278 | |
| RDULEYWUGKOCMR-UHFFFAOYNA-N | |
| ethyl 2-chloro-3-oxobutanoate | |
| 11858 | |
| 96% |
Chemical Identifiers
| 609-15-4 | |
| 164.59 | |
| RDULEYWUGKOCMR-UHFFFAOYNA-N | |
| ethyl 2-chloro-3-oxobutanoate |
| C6H9ClO3 | |
| MFCD00009141 | |
| 11858 | |
| CCOC(=O)C(Cl)C(C)=O |
Safety and Handling
GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 210-180-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only