missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Chloro-6-fluorophenylacetate 98.0+%, TCI America™

Catalog No. E10381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10381G 1 g
E10385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10381G Supplier TCI America Supplier No. E10381G
Only null left

Chemical Identifiers

CAS 214262-85-8
Molecular Formula C10H10ClFO2
Molecular Weight (g/mol) 216.636
MDL Number MFCD00800606
InChI Key CEWCLHPMDWPDNQ-UHFFFAOYSA-N
PubChem CID 2733231
IUPAC Name ethyl 2-(2-chloro-6-fluorophenyl)acetate
SMILES CCOC(=O)CC1=C(C=CC=C1Cl)F

Specifications

Color Yellow
Quantity 1 g
Synonym ethyl 2-chloro-6-fluorophenylacetate, ethyl 2-2-chloro-6-fluorophenyl acetate, 2-chloro-6-fluorophenyl acetic acid ethyl ester, ethyl2-chloro-6-fluorophenylacetate, ethyl 2-chloro-6-fluorophenyl acetate, ethyl 2-2-chloro-6-fluoro-phenyl acetate, 2-chloro-6-fluorophenylacetic acid ethyl ester, 2-chloro-6-fluoro-phenyl-acetic acid ethyl ester, benzeneacetic acid,2-chloro-6-fluoro-, ethyl ester
Formula Weight 216.64
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Chloro-6-fluorophenylacetate