missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-chloro-5-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
$169.30 - $169.30
Chemical Identifiers
| CAS | 902837-55-2 |
|---|---|
| Molecular Formula | C9H8ClNO3 |
| Molecular Weight (g/mol) | 213.617 |
| MDL Number | MFCD07781188 |
| InChI Key | HRFPVENOVYSKHE-UHFFFAOYSA-N |
| Synonym | ethyl 6-chloro-3-pyridylglyoxylate, ethyl 2-6-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloro-5-pyridylglyoxylate, ethyl 6-chloropyridin-3-yl oxo acetate, ethyl6-chloro-3-pyridylglyoxylate, ethyl 6-chloro-3-pyridinyl oxoacetate, 6-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-6-chloro 3-pyridyl-2-oxoacetate |
| PubChem CID | 42553168 |
| IUPAC Name | ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate |
| SMILES | CCOC(=O)C(=O)C1=CN=C(C=C1)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5015703
|
Thermo Scientific Chemicals
H5015703 |
1 g |
Each for $169.30
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 902837-55-2 | |
| 213.617 | |
| HRFPVENOVYSKHE-UHFFFAOYSA-N | |
| 42553168 | |
| CCOC(=O)C(=O)C1=CN=C(C=C1)Cl |
| C9H8ClNO3 | |
| MFCD07781188 | |
| ethyl 6-chloro-3-pyridylglyoxylate, ethyl 2-6-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloro-5-pyridylglyoxylate, ethyl 6-chloropyridin-3-yl oxo acetate, ethyl6-chloro-3-pyridylglyoxylate, ethyl 6-chloro-3-pyridinyl oxoacetate, 6-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-6-chloro 3-pyridyl-2-oxoacetate | |
| ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate |
Specifications
| 902837-55-2 | |
| MFCD07781188 | |
| ethyl 6-chloro-3-pyridylglyoxylate, ethyl 2-6-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloro-5-pyridylglyoxylate, ethyl 6-chloropyridin-3-yl oxo acetate, ethyl6-chloro-3-pyridylglyoxylate, ethyl 6-chloro-3-pyridinyl oxoacetate, 6-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-6-chloro 3-pyridyl-2-oxoacetate | |
| CCOC(=O)C(=O)C1=CN=C(C=C1)Cl | |
| 213.617 | |
| 213.62 | |
| Ethyl 2-chloro-5-pyridylglyoxylate |
| C9H8ClNO3 | |
| 1 g | |
| HRFPVENOVYSKHE-UHFFFAOYSA-N | |
| ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate | |
| 42553168 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only