accessibility menu, dialog, popup

UserName

CAS RN 902837-55-2

H3COOONCl

CAS RN 902837-55-2

IUPAC Name: ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate
Synonyms: ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate
Molecular Weight (g/mol): 213.62
Molecular Formula: C9H8ClNO3
InChi Key: HRFPVENOVYSKHE-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=CN=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results