missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate, 95%

Catalog No. aah6115803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6115803 1 g
AAH6115806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6115803 Supplier Thermo Scientific Chemicals Supplier No. H6115803
Only null left

CAS: 188781-08-0 | C8H9ClN2O2 | 200.622 g/mol

It is an active pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool dry ventilated place. Ensure proper ventilation.

Chemical Identifiers

CAS 188781-08-0
Molecular Formula C8H9ClN2O2
Molecular Weight (g/mol) 200.622
MDL Number MFCD03425963
InChI Key XJNJPQSXRUJCIB-UHFFFAOYSA-N
Synonym 2-chloro-4-methyl-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-5-ethoxycarbonyl-4-methylpyrimidine, ethyl2-chloro-4-methylpyrimidine-5-carboxylate, 2-chloro-4-methyl-5-pyrimidinecarboxylic acid ethyl ester
PubChem CID 21220624
IUPAC Name ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1C)Cl

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 200.62
Percent Purity 95%
Chemical Name or Material Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only