CAS RN 188781-08-0
IUPAC Name:
ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
Synonyms:
ethyl 2-chloro-4-methylpyrimidine-5-carboxylate
Molecular Weight (g/mol):
200.62
Molecular Formula:
C8H9ClN2O2
InChi Key:
XJNJPQSXRUJCIB-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CN=C(Cl)N=C1C
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate, 95%
Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate, 95%
CAS: 188781-08-0 Molecular Formula: C8H9ClN2O2 Molecular Weight (g/mol): 200.622 MDL Number: MFCD03425963 InChI Key: XJNJPQSXRUJCIB-UHFFFAOYSA-N Synonym: 2-chloro-4-methyl-pyrimidine-5-carboxylic acid ethyl ester,5-pyrimidinecarboxylic acid, 2-chloro-4-methyl-, ethyl ester,2-chloro-5-ethoxycarbonyl-4-methylpyrimidine,ethyl2-chloro-4-methylpyrimidine-5-carboxylate,2-chloro-4-methyl-5-pyrimidinecarboxylic acid ethyl ester PubChem CID: 21220624 IUPAC Name: ethyl 2-chloro-4-methylpyrimidine-5-carboxylate SMILES: CCOC(=O)C1=CN=C(N=C1C)Cl