missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Butynoate 98.0+%, TCI America™
Supplier: TCI America T10521ML
Specifications
| Ethyl 2-Butynoate | |
| Yellow | |
| C6H8O2 | |
| 1 mL | |
| FCJJZKCJURDYNF-UHFFFAOYSA-N | |
| ethyl but-2-ynoate | |
| 78043 | |
| ≥98.0% (GC) |
| 4341-76-8 | |
| 163°C | |
| MFCD00015182 | |
| ethyl 2-butynoate, ethyl tetrolate, 2-butynoic acid ethyl ester, but-2-ynoic acid ethyl ester, 2-butynoic acid, ethyl ester, tetrolic acid ethyl ester, ethyl-2-butynoate, ethyl 3-methylpropiolate, acmc-1aeo2, ethyl 1-propynecarboxylate | |
| CCOC(=O)C#CC | |
| 112.128 | |
| 112.13 | |
| Liquid |
Chemical Identifiers
| 4341-76-8 | |
| 112.128 | |
| FCJJZKCJURDYNF-UHFFFAOYSA-N | |
| 78043 | |
| CCOC(=O)C#CC |
| C6H8O2 | |
| MFCD00015182 | |
| ethyl 2-butynoate, ethyl tetrolate, 2-butynoic acid ethyl ester, but-2-ynoic acid ethyl ester, 2-butynoic acid, ethyl ester, tetrolic acid ethyl ester, ethyl-2-butynoate, ethyl 3-methylpropiolate, acmc-1aeo2, ethyl 1-propynecarboxylate | |
| ethyl but-2-ynoate |
Safety and Handling
TSCA : No