missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Butynoate 98.0+%, TCI America™

Catalog No. p-7136576
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10521ML 1 mL
T10525ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10521ML Supplier TCI America Supplier No. T10521ML

Chemical Identifiers

CAS 4341-76-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00015182
InChI Key FCJJZKCJURDYNF-UHFFFAOYSA-N
Synonym ethyl 2-butynoate, ethyl tetrolate, 2-butynoic acid ethyl ester, but-2-ynoic acid ethyl ester, 2-butynoic acid, ethyl ester, tetrolic acid ethyl ester, ethyl-2-butynoate, ethyl 3-methylpropiolate, acmc-1aeo2, ethyl 1-propynecarboxylate
PubChem CID 78043
IUPAC Name ethyl but-2-ynoate
SMILES CCOC(=O)C#CC

Specifications

Color Yellow
Boiling Point 163°C
Quantity 1 mL
Formula Weight 112.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Butynoate