missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromothiazole-4-carboxylate 95.0+%, TCI America™

Catalog No. E09001G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09001G 1 g
E09005G 5 g
E090025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E09001G Supplier TCI America Supplier No. E09001G

Chemical Identifiers

CAS 100367-77-9
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.083
MDL Number MFCD03788564
InChI Key CNHISCQPKKGDPO-UHFFFAOYSA-N
PubChem CID 353965
IUPAC Name ethyl 2-bromo-1,3-thiazole-4-carboxylate
SMILES CCOC(=O)C1=CSC(=N1)Br

Specifications

Melting Point 70°C
Color White-Yellow
Boiling Point 154°C
Quantity 1 g
Synonym ethyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid ethyl ester, 4-thiazolecarboxylic acid, 2-bromo-, ethyl ester, ethyl2-bromothiazole-4-carboxylate, ethyl 2-bromo-4-thiazolecarboxylate, 2-bromo-thiazole-4-carboxylic acid ethyl ester, 2-bromo-4-ethoxycarbonyl-1,3-thiazole, 2-bromo-1,3-thiazole-4-carboxylic acid ethyl ester, zlchem 490, pubchem17466
Formula Weight 236.08
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Bromothiazole-4-carboxylate
TSCA No
Recommended Storage Refrigerator