accessibility menu, dialog, popup

UserName

CAS RN 100367-77-9

IUPAC Name: ethyl 2-bromo-1,3-thiazole-4-carboxylate
Synonyms: ethyl 2-bromo-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 236.08
Molecular Formula: C6H6BrNO2S
InChi Key: CNHISCQPKKGDPO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CSC(Br)=N1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)