Learn More
Ethyl 2-bromopropionate, 98+%
CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol
Supplier: Thermo Scientific Chemicals A1082030
Description
Itâs an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsItâs an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.
Solubility
Immiscible in water. Soluble in alcohol and ether.
Notes
Store in dark and away from oxidizing agents and acids.
Specifications
| Ethyl 2-bromopropionate | |
| 1.391 | |
| 51°C (123°F) | |
| 1.446 | |
| 250 g | |
| 773920 | |
| ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate | |
| ARFLASKVLJTEJD-UHFFFAOYNA-N | |
| ethyl 2-bromopropanoate | |
| 79040 | |
| ≥98% |
| 535-11-5 | |
| 158°C to 160°C | |
| C5H9BrO2 | |
| MFCD00000144 | |
| UN1992 | |
| 14,3772 | |
| Immiscible in water. Soluble in alcohol and ether. | |
| CCOC(=O)C(C)Br | |
| 181.03 | |
| 181.03 |
Chemical Identifiers
| 535-11-5 | |
| 181.03 | |
| ARFLASKVLJTEJD-UHFFFAOYNA-N | |
| 79040 | |
| CCOC(=O)C(C)Br |
| C5H9BrO2 | |
| MFCD00000144 | |
| ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate | |
| ethyl 2-bromopropanoate |
Safety and Handling
GHS H Statement
H318-H226-H302-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 208-609-5
RTECSNumber : UA2451730
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only