missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(Bromomethyl)acrylate (stabilized with HQ) 97.0+%, TCI America™

Catalog No. p-7119285
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18201G 1 g
B18205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18201G Supplier TCI America Supplier No. B18201G

Chemical Identifiers

CAS 17435-72-2
Molecular Formula C6H9BrO2
Molecular Weight (g/mol) 193.04
MDL Number MFCD00031518
InChI Key MTCMFVTVXAOHNQ-UHFFFAOYSA-N
Synonym ethyl 2-bromomethyl acrylate, ethyl 2-bromomethyl prop-2-enoate, 2-bromomethyl acrylic acid ethyl ester, ethyl alpha-bromomethyl acrylate, 2-propenoic acid, 2-bromomethyl-, ethyl ester, 2-bromomethyl-acrylic acid ethyl ester, ethyl-2-bromomethylacrylate, ethyl 2-bromomethylacrylate, acmc-1bo78, ksc181c2b
PubChem CID 310620
IUPAC Name ethyl 2-(bromomethyl)prop-2-enoate
SMILES CCOC(=O)C(=C)CBr

Specifications

Color Yellow
Boiling Point 86°C
Quantity 1 g
Formula Weight 193.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-(Bromomethyl)acrylate (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator