accessibility menu, dialog, popup

UserName

H3COOCH2Br

CAS RN 17435-72-2

IUPAC Name: ethyl 2-(bromomethyl)prop-2-enoate
Synonyms: ethyl 2-(bromomethyl)prop-2-enoate
Molecular Weight (g/mol): 193.04
Molecular Formula: C6H9BrO2
InChi Key: MTCMFVTVXAOHNQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=C)CBr
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (6)
  • (3)
  • (3)