missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(bromomethyl)acrylate, 97%

Catalog No. AC235360050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC235360050 5 g Glass bottle
AC235360010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC235360050 Supplier Thermo Scientific Chemicals Supplier No. 235360050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17435-72-2 | C6H9BrO2 | 193.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17435-72-2
Molecular Formula C6H9BrO2
Molecular Weight (g/mol) 193.04
MDL Number MFCD00031518
InChI Key MTCMFVTVXAOHNQ-UHFFFAOYSA-N
Synonym ethyl 2-bromomethyl acrylate, ethyl 2-bromomethyl prop-2-enoate, 2-bromomethyl acrylic acid ethyl ester, ethyl alpha-bromomethyl acrylate, 2-propenoic acid, 2-bromomethyl-, ethyl ester, 2-bromomethyl-acrylic acid ethyl ester, ethyl-2-bromomethylacrylate, ethyl 2-bromomethylacrylate, acmc-1bo78, ksc181c2b
PubChem CID 310620
IUPAC Name ethyl 2-(bromomethyl)prop-2-enoate
SMILES CCOC(=O)C(=C)CBr

Specifications

Density 1.3800g/mL
Boiling Point 86°C (20.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4770 to 1.479
Linear Formula CH2=C(CH2Br)COOC2H5
Quantity 5 g
Fieser 13,322; 14,162
Specific Gravity 1.38
Formula Weight 193.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-(bromomethyl)acrylate, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only