missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromoisobutyrate 98.0+%, TCI America™

Catalog No. p-7118561
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B060625G 25 g
B0606100G 100 g
B0606500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B060625G Supplier TCI America Supplier No. B060625G
Only null left

Chemical Identifiers

CAS 600-00-0
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000123
InChI Key IOLQWGVDEFWYNP-UHFFFAOYSA-N
Synonym ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester
PubChem CID 11745
IUPAC Name ethyl 2-bromo-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)Br

Specifications

Color Yellow
Boiling Point 161°C
Quantity 25 g
UN Number 2920
Formula Weight 195.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Bromoisobutyrate
EINECSNumber (2)-1179&(2)-2650
TSCA Yes