missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%

Catalog No. p-7031480
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32572MD 250 mg
AAH3257203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32572MD Supplier Thermo Scientific Chemicals Supplier No. H32572MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 367-33-9 | C6H8BrF3O2 | 249.027 g/mol

Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 367-33-9
Molecular Formula C6H8BrF3O2
Molecular Weight (g/mol) 249.027
MDL Number MFCD10687235
InChI Key POVYUFTVLGOXEE-UHFFFAOYSA-N
Synonym 2-bromo-4,4,4-trifluorobutyric acid ethyl ester, ethyl 2-bromo-4,4,4-trifluorobutyrate, ethyl 2-bromo-4,4,4-trifluoro-butanoate, butanoic acid,2-bromo-4,4,4-trifluoro-, ethyl ester
PubChem CID 45594306
IUPAC Name ethyl 2-bromo-4,4,4-trifluorobutanoate
SMILES CCOC(=O)C(CC(F)(F)F)Br

Specifications

Boiling Point 67°C (18 mmHg)
Quantity 250 mg
UN Number UN3265
Formula Weight 249.03
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromo-4,4,4-trifluorobutyrate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only