missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Benzylacetoacetate 97.0+%, TCI America™

Catalog No. p-7119417
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20085G 5 g
B200825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20085G Supplier TCI America Supplier No. B20085G

Chemical Identifiers

CAS 620-79-1
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
MDL Number MFCD00009156
InChI Key XDWQYMXQMNUWID-UHFFFAOYSA-N
Synonym ethyl 2-benzylacetoacetate, ethyl 2-acetyl-3-phenylpropionate, ethyl benzyl acetoacetate, ethyl alpha-benzylacetoacetate, 2-benzylacetoacetic acid ethyl ester, ethyl .alpha.-benzylacetoacetate, 2-benzylacetoacetate, ethyl 3-oxo-2-benzylbutanoate, ethyl alpha-acetylhydrocinnamate, ethyl benzylacetoacetate
PubChem CID 246929
IUPAC Name ethyl 2-benzyl-3-oxobutanoate
SMILES CCOC(=O)C(CC1=CC=CC=C1)C(=O)C

Specifications

Color Yellow
Boiling Point 284°C
Quantity 5 g
Formula Weight 220.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Benzylacetoacetate
EINECSNumber (3)-3090
TSCA No