accessibility menu, dialog, popup

UserName

H3COOCH3O

CAS RN 620-79-1

IUPAC Name: ethyl 2-benzyl-3-oxobutanoate
Synonyms: ethyl 2-benzyl-3-oxobutanoate
Molecular Weight (g/mol): 220.27
Molecular Formula: C13H16O3
InChi Key: XDWQYMXQMNUWID-UHFFFAOYNA-N
SMILES: CCOC(=O)C(CC1=CC=CC=C1)C(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results