missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminothiophene-3-carboxylate, 97%

Catalog No. AAH6155922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6155922 100 g
AAH6155914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6155922 Supplier Thermo Scientific Chemicals Supplier No. H6155922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31891-06-2 | C7H9NO2S | 171.214 g/mol

Ethyl 2-aminothiophene-3-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-aminothiophene-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 31891-06-2
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD01566303
InChI Key MKJQYFVTEPGXIE-UHFFFAOYSA-N
Synonym 2-amino-thiophene-3-carboxylic acid ethyl ester, 2-aminothiophene-3-carboxylic acid ethyl ester, ethyl2-aminothiophene-3-carboxylate, 2-amino-3-ethoxycarbonylthiophene, ethyl 2-amino-3-thiophenecarboxylate, 2-aminothiophene-3-carboxylate, 3-thiophenecarboxylic acid, 2-amino-, ethyl ester, pubchem22914, acmc-209hpg, ethyl 2-amino-3-thenoate
PubChem CID 1988156
IUPAC Name ethyl 2-aminothiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1)N

Specifications

Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 171.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 2-aminothiophene-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only