CAS RN 31891-06-2
IUPAC Name:
ethyl 2-aminothiophene-3-carboxylate
Synonyms:
ethyl 2-aminothiophene-3-carboxylate
Molecular Weight (g/mol):
171.21
Molecular Formula:
C7H9NO2S
InChi Key:
MKJQYFVTEPGXIE-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(N)SC=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-aminothiophene-3-carboxylate, 97%
Ethyl 2-aminothiophene-3-carboxylate, 97%
CAS: 31891-06-2 Molecular Formula: C7H9NO2S Molecular Weight (g/mol): 171.214 MDL Number: MFCD01566303 InChI Key: MKJQYFVTEPGXIE-UHFFFAOYSA-N Synonym: 2-amino-thiophene-3-carboxylic acid ethyl ester,2-aminothiophene-3-carboxylic acid ethyl ester,ethyl2-aminothiophene-3-carboxylate,2-amino-3-ethoxycarbonylthiophene,ethyl 2-amino-3-thiophenecarboxylate,2-aminothiophene-3-carboxylate,3-thiophenecarboxylic acid, 2-amino-, ethyl ester,pubchem22914,acmc-209hpg,ethyl 2-amino-3-thenoate PubChem CID: 1988156 IUPAC Name: ethyl 2-aminothiophene-3-carboxylate SMILES: CCOC(=O)C1=C(SC=C1)N