missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Aminothiazole-5-carboxylate 97.0+%, TCI America™
$61.51 - $70.84
Chemical Identifiers
| CAS | 32955-21-8 |
|---|---|
| Molecular Formula | C6H8N2O2S |
| Molecular Weight (g/mol) | 172.202 |
| MDL Number | MFCD00602139 |
| InChI Key | VNZXERIGKZNEKB-UHFFFAOYSA-N |
| Synonym | ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456 |
| PubChem CID | 314628 |
| IUPAC Name | ethyl 2-amino-1,3-thiazole-5-carboxylate |
| SMILES | CCOC(=O)C1=CN=C(S1)N |
Chemical Identifiers
| 32955-21-8 | |
| 172.202 | |
| VNZXERIGKZNEKB-UHFFFAOYSA-N | |
| 314628 | |
| CCOC(=O)C1=CN=C(S1)N |
| C6H8N2O2S | |
| MFCD00602139 | |
| ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456 | |
| ethyl 2-amino-1,3-thiazole-5-carboxylate |
Specifications
| 32955-21-8 | |
| C6H8N2O2S | |
| 1 g | |
| VNZXERIGKZNEKB-UHFFFAOYSA-N | |
| ethyl 2-amino-1,3-thiazole-5-carboxylate | |
| 314628 | |
| ≥97.0% (HPLC,T) | |
| Ethyl 2-Aminothiazole-5-carboxylate |
| 161°C | |
| MFCD00602139 | |
| ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456 | |
| CCOC(=O)C1=CN=C(S1)N | |
| 172.202 | |
| 172.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator