accessibility menu, dialog, popup

UserName

CAS RN 32955-21-8

IUPAC Name: ethyl 2-amino-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-amino-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: VNZXERIGKZNEKB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(N)S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)