missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Aminothiazole-5-carboxylate 97.0+%, TCI America™

Catalog No. E09071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09071G 1 g
E09075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09071G Supplier TCI America Supplier No. E09071G

Chemical Identifiers

CAS 32955-21-8
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00602139
InChI Key VNZXERIGKZNEKB-UHFFFAOYSA-N
Synonym ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456
PubChem CID 314628
IUPAC Name ethyl 2-amino-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=CN=C(S1)N

Specifications

Melting Point 161°C
Quantity 1 g
Formula Weight 172.20
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Aminothiazole-5-carboxylate
TSCA No
Recommended Storage Refrigerator