missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-aminothiazole-5-carboxylate, 97%
CAS: 32955-21-8 | C6H8N2O2S | 172.202 g/mol
$74.51 - $248.90
Chemical Identifiers
| CAS | 32955-21-8 |
|---|---|
| Molecular Formula | C6H8N2O2S |
| Molecular Weight (g/mol) | 172.202 |
| MDL Number | MFCD00602139 |
| InChI Key | VNZXERIGKZNEKB-UHFFFAOYSA-N |
| Synonym | ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456 |
| PubChem CID | 314628 |
| IUPAC Name | ethyl 2-amino-1,3-thiazole-5-carboxylate |
| SMILES | CCOC(=O)C1=CN=C(S1)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3176903
|
Thermo Scientific Chemicals
H3176903 |
1 g |
Each for $74.51
|
|
|||||
|
AAH3176906
|
Thermo Scientific Chemicals
H3176906 |
5 g |
Each for $248.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 32955-21-8 | |
| 172.202 | |
| VNZXERIGKZNEKB-UHFFFAOYSA-N | |
| 314628 | |
| CCOC(=O)C1=CN=C(S1)N |
| C6H8N2O2S | |
| MFCD00602139 | |
| ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456 | |
| ethyl 2-amino-1,3-thiazole-5-carboxylate |
Specifications
| 32955-21-8 | |
| C6H8N2O2S | |
| 1 g | |
| ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456 | |
| CCOC(=O)C1=CN=C(S1)N | |
| 172.202 | |
| 172.21 | |
| Ethyl 2-aminothiazole-5-carboxylate |
| 158°C to 160°C | |
| MFCD00602139 | |
| Light and moisture sensitive | |
| VNZXERIGKZNEKB-UHFFFAOYSA-N | |
| ethyl 2-amino-1,3-thiazole-5-carboxylate | |
| 314628 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only