missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminothiazole-5-carboxylate, 97%

Catalog No. AAH3176906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3176906 5 g
AAH3176903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3176906 Supplier Thermo Scientific Chemicals Supplier No. H3176906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32955-21-8 | C6H8N2O2S | 172.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32955-21-8
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00602139
InChI Key VNZXERIGKZNEKB-UHFFFAOYSA-N
Synonym ethyl 2-aminothiazole-5-carboxylate, ethyl 2-amino-thiazole-5-carboxylate, 2-amino-thiazole-5-carboxylic acid ethyl ester, ethyl2-aminothiazole-5-carboxylate, 2-amino-thiazole-5-carboxylate ethyl ester, 2-aminothiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-5-thiazolecarboxylic acid ethyl ester, ethyl-2-amino thiazole-5-carboxylate, zlchem 456
PubChem CID 314628
IUPAC Name ethyl 2-amino-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=CN=C(S1)N

Specifications

Melting Point 158°C to 160°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Formula Weight 172.21
Percent Purity 97%
Chemical Name or Material Ethyl 2-aminothiazole-5-carboxylate
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only