missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Aminothiazole-4-carboxylate 98.0+%, TCI America™
$67.38 - $200.21
Chemical Identifiers
| CAS | 5398-36-7 |
|---|---|
| Molecular Formula | C6H8N2O2S |
| Molecular Weight (g/mol) | 172.202 |
| MDL Number | MFCD00619079 |
| InChI Key | XHFUVBWCMLLKOZ-UHFFFAOYSA-N |
| Synonym | ethyl 2-aminothiazole-4-carboxylate, 2-amino-thiazole-4-carboxylic acid ethyl ester, ethyl 2-amino-4-thiazolecarboxylate, 4-thiazolecarboxylic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazoleformate, 2-amino-4-ethoxycarbonylthiazole, 2-aminothiazole-4-ethyl formate, 2-aminothiazole-4-carboxylic acid ethyl ester, ethyl-2-aminothiazole-4-carboxylate, ethyl-2-aminothiazole-4-carboxylic acid |
| PubChem CID | 73216 |
| IUPAC Name | ethyl 2-amino-1,3-thiazole-4-carboxylate |
| SMILES | CCOC(=O)C1=CSC(=N1)N |
Chemical Identifiers
| 5398-36-7 | |
| 172.202 | |
| XHFUVBWCMLLKOZ-UHFFFAOYSA-N | |
| 73216 | |
| CCOC(=O)C1=CSC(=N1)N |
| C6H8N2O2S | |
| MFCD00619079 | |
| ethyl 2-aminothiazole-4-carboxylate, 2-amino-thiazole-4-carboxylic acid ethyl ester, ethyl 2-amino-4-thiazolecarboxylate, 4-thiazolecarboxylic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazoleformate, 2-amino-4-ethoxycarbonylthiazole, 2-aminothiazole-4-ethyl formate, 2-aminothiazole-4-carboxylic acid ethyl ester, ethyl-2-aminothiazole-4-carboxylate, ethyl-2-aminothiazole-4-carboxylic acid | |
| ethyl 2-amino-1,3-thiazole-4-carboxylate |
Specifications
| 5398-36-7 | |
| White-Yellow | |
| MFCD00619079 | |
| ethyl 2-aminothiazole-4-carboxylate, 2-amino-thiazole-4-carboxylic acid ethyl ester, ethyl 2-amino-4-thiazolecarboxylate, 4-thiazolecarboxylic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazoleformate, 2-amino-4-ethoxycarbonylthiazole, 2-aminothiazole-4-ethyl formate, 2-aminothiazole-4-carboxylic acid ethyl ester, ethyl-2-aminothiazole-4-carboxylate, ethyl-2-aminothiazole-4-carboxylic acid | |
| CCOC(=O)C1=CSC(=N1)N | |
| 172.202 | |
| 172.20 | |
| Crystalline Powder |
| 180°C | |
| C6H8N2O2S | |
| 5 g | |
| XHFUVBWCMLLKOZ-UHFFFAOYSA-N | |
| ethyl 2-amino-1,3-thiazole-4-carboxylate | |
| 73216 | |
| ≥98.0% (GC,T) | |
| Ethyl 2-Aminothiazole-4-carboxylate |
Safety and Handling
TSCA : No