accessibility menu, dialog, popup

UserName

CH3OOSH2NN

CAS RN 5398-36-7

IUPAC Name: ethyl 2-amino-1,3-thiazole-4-carboxylate
Synonyms: ethyl 2-amino-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: XHFUVBWCMLLKOZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CSC(N)=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)