missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Aminothiazole-4-carboxylate 98.0+%, TCI America™

Catalog No. E12155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12155G 5 g
E121525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12155G Supplier TCI America Supplier No. E12155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5398-36-7
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00619079
InChI Key XHFUVBWCMLLKOZ-UHFFFAOYSA-N
Synonym ethyl 2-aminothiazole-4-carboxylate, 2-amino-thiazole-4-carboxylic acid ethyl ester, ethyl 2-amino-4-thiazolecarboxylate, 4-thiazolecarboxylic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazoleformate, 2-amino-4-ethoxycarbonylthiazole, 2-aminothiazole-4-ethyl formate, 2-aminothiazole-4-carboxylic acid ethyl ester, ethyl-2-aminothiazole-4-carboxylate, ethyl-2-aminothiazole-4-carboxylic acid
PubChem CID 73216
IUPAC Name ethyl 2-amino-1,3-thiazole-4-carboxylate
SMILES CCOC(=O)C1=CSC(=N1)N

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
Formula Weight 172.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Aminothiazole-4-carboxylate