missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminooxazole-4-carboxylate, 95%

Catalog No. p-7031457
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3254203 1 g
AAH3254206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3254203 Supplier Thermo Scientific Chemicals Supplier No. H3254203
Only null left

CAS: 177760-52-0 | C6H8N2O3 | 156.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 177760-52-0
Molecular Formula C6H8N2O3
Molecular Weight (g/mol) 156.14
MDL Number MFCD06659907
InChI Key NBABLVASYFPOEV-UHFFFAOYSA-N
PubChem CID 319549
IUPAC Name ethyl 2-amino-1,3-oxazole-4-carboxylate
SMILES CCOC(=O)C1=COC(N)=N1

Specifications

Melting Point 124°C to 128°C
Quantity 1 g
Synonym ethyl 2-aminooxazole-4-carboxylate, 2-aminooxazole-4-carboxylic acid ethyl ester, ethyl2-aminooxazole-4-carboxylate, 2-amino-oxazole-4-carboxylic acid ethyl ester, 4-oxazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-4-ethoxycarbonyl-1,3-oxazole, ethyl 2-amino-4-oxazolecarboxylate, pubchem18558, acmc-1c3gt
Formula Weight 156.14
Percent Purity 95%
Chemical Name or Material Ethyl 2-aminooxazole-4-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only