CAS RN 177760-52-0
IUPAC Name:
ethyl 2-amino-1,3-oxazole-4-carboxylate
Synonyms:
ethyl 2-amino-1,3-oxazole-4-carboxylate
Molecular Weight (g/mol):
156.14
Molecular Formula:
C6H8N2O3
InChi Key:
NBABLVASYFPOEV-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=COC(N)=N1
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-aminooxazole-4-carboxylate, 95%
Ethyl 2-aminooxazole-4-carboxylate, 95%
CAS: 177760-52-0 Molecular Formula: C6H8N2O3 Molecular Weight (g/mol): 156.14 MDL Number: MFCD06659907 InChI Key: NBABLVASYFPOEV-UHFFFAOYSA-N PubChem CID: 319549 IUPAC Name: ethyl 2-amino-1,3-oxazole-4-carboxylate SMILES: CCOC(=O)C1=COC(N)=N1