missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Aminonicotinate 98.0+%, TCI America™

Catalog No. p-7129359
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11461G 1 g
E11465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11461G Supplier TCI America Supplier No. E11461G
Only null left

Chemical Identifiers

CAS 13362-26-0
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD01569452
InChI Key KIAGEDYOPMHRRB-UHFFFAOYSA-N
Synonym ethyl 2-aminonicotinate, 2-amino-nicotinic acid ethyl ester, ethyl2-aminonicotinate, 2-aminonicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino nicotinic acid ethyl ester, 2-aminopyridine-3-carboxylic acid ethyl ester, pubchem12921, rarechem al bi 1234, buttpark 23\07-65
PubChem CID 12433260
IUPAC Name ethyl 2-aminopyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)N

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 133°C
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Aminonicotinate