accessibility menu, dialog, popup

UserName

H3COONNH2

CAS RN 13362-26-0

IUPAC Name: ethyl 2-aminopyridine-3-carboxylate
Synonyms: ethyl 2-aminopyridine-3-carboxylate
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: KIAGEDYOPMHRRB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)N=CC=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)