missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminonicotinate, 98%

Catalog No. p-7056182
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6495706 5 g
AAH6495714 25 g
AAH6495722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6495706 Supplier Thermo Scientific Chemicals Supplier No. H6495706

CAS: 13362-26-0 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13362-26-0
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD01569452
InChI Key KIAGEDYOPMHRRB-UHFFFAOYSA-N
PubChem CID 12433260
IUPAC Name ethyl 2-aminopyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)N

Specifications

Melting Point 90°C to 96°C (literature)
Quantity 5 g
Beilstein 124680
Synonym ethyl 2-aminonicotinate, 2-amino-nicotinic acid ethyl ester, ethyl2-aminonicotinate, 2-aminonicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino nicotinic acid ethyl ester, 2-aminopyridine-3-carboxylic acid ethyl ester, pubchem12921, rarechem al bi 1234, buttpark 23\07-65
Formula Weight 166.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 2-aminonicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only