missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-5-phenylthiophene-3-carboxylate 98.0+%, TCI America™

Catalog No. p-7129355
Change view
Click to view available options
Quantity:
1 g
200 mg
Catalog No. Quantity
E1139200MG 200 mg
E11391G 1 g
2 options

Catalog No. E1139200MG

Supplier: TCI America E1139200MG

Only null left

Chemical Identifiers

CAS 4815-34-3
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
MDL Number MFCD01829801
InChI Key WIVNPGXPJBBZQH-UHFFFAOYSA-N
Synonym ethyl 2-amino-5-phenyl-3-thiophenecarboxylate, 2-amino-5-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-5-phenylthiophene-3-carboxylic acid ethyl ester, ethyl2-amino-5-phenylthiophene-3-carboxylate, 2-amino-5-phenyl-3-thiophenecarboxylic acid ethyl ester, enamine_004942, 2-amino-3-ethoxycarbonyl-5-phenylthiophene, ethyl 2-azanyl-5-phenyl-thiophene-3-carboxylate, 2-amino-5-phenyl thiophene-3-carboxylic ethyl ester, 2-amino-5-phenyl-3-thiophenecarboxylic acid, ethyl ester
PubChem CID 638860
IUPAC Name ethyl 2-amino-5-phenylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1)C2=CC=CC=C2)N

Specifications

Melting Point 124°C
Color Yellow
Quantity 200 mg
Formula Weight 247.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-5-phenylthiophene-3-carboxylate