missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-5-phenylthiophene-3-carboxylate 98.0+%, TCI America™

Catalog No. E11391G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11391G 1 g
E1139200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11391G Supplier TCI America Supplier No. E11391G

Chemical Identifiers

CAS 4815-34-3
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
MDL Number MFCD01829801
InChI Key WIVNPGXPJBBZQH-UHFFFAOYSA-N
Synonym ethyl 2-amino-5-phenyl-3-thiophenecarboxylate, 2-amino-5-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-5-phenylthiophene-3-carboxylic acid ethyl ester, ethyl2-amino-5-phenylthiophene-3-carboxylate, 2-amino-5-phenyl-3-thiophenecarboxylic acid ethyl ester, enamine_004942, 2-amino-3-ethoxycarbonyl-5-phenylthiophene, ethyl 2-azanyl-5-phenyl-thiophene-3-carboxylate, 2-amino-5-phenyl thiophene-3-carboxylic ethyl ester, 2-amino-5-phenyl-3-thiophenecarboxylic acid, ethyl ester
PubChem CID 638860
IUPAC Name ethyl 2-amino-5-phenylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1)C2=CC=CC=C2)N

Specifications

Melting Point 124°C
Color Yellow
Quantity 1 g
Formula Weight 247.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-5-phenylthiophene-3-carboxylate