missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-methylthiophene-3-carboxylate, 98+%

Catalog No. AAA1079006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1079006 5 g
AAA1079014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1079006 Supplier Thermo Scientific Chemicals Supplier No. A1079006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 43088-42-2 | C8H11NO2S | 185.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 43088-42-2
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD00051669
InChI Key ILYCZKOBLRJJSW-UHFFFAOYSA-N
Synonym 2-amino-4-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-4-methyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl2-amino-4-methylthiophene-3-carboxylate, ethyl 2-amino-4-methyl-3-thiophenecarboxylate, acmc-20a9ux, maybridge1_003755, 2-amino-4-methyl-3-ethoxycarbonyl-thiophene, ethyl 2-amino4-methylthiophene-3-carboxylate, 2-amino-3-ethoxycarbonyl-4-methylthiophene
PubChem CID 521132
IUPAC Name ethyl 2-amino-4-methylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C)N

Specifications

Melting Point 79°C to 83°C
Quantity 5 g
Beilstein 1366073
Formula Weight 185.25
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-amino-4-methylthiophene-3-carboxylate
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only