missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate, 95%

Catalog No. AAL0665403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0665403 1 g
AAL0665406 5 g
AAL0665414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0665403 Supplier Thermo Scientific Chemicals Supplier No. L0665403

CAS: 15400-53-0 | C7H9N3O3 | 183.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15400-53-0
Molecular Formula C7H9N3O3
Molecular Weight (g/mol) 183.167
MDL Number MFCD00039708
InChI Key HRRHGLKNOJHIGY-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate, 2-amino-5-carboethoxy-4-hydroxypyrimidine, 2-amino-4-hydroxypyrimidine-5-carboxylic acid ethyl ester, ethyl 2-amino-1,4-dihydro-4-oxopyrimidine-5-carboxylate, 2-amino-4-hydroxy-pyrimidine-5-carboxylic acid ethyl ester, 2-amino-6-oxo-1,6-dihydro-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 2-amino-1,4-dihydro-4-oxo-, ethyl ester, ethyl 2-amino-4-oxo-3h-pyrimidine-5-carboxylate, 1rry, acmc-209xd5
PubChem CID 84894
ChEBI CHEBI:39721
IUPAC Name ethyl 2-amino-6-oxo-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(NC1=O)N

Specifications

Melting Point ∽300°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 165835
Formula Weight 183.17
Percent Purity 95%
Chemical Name or Material Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-418-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only