missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-acetylisonicotinate, 96%

Catalog No. p-7052568
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3691103 1 g
AAH3691106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3691103 Supplier Thermo Scientific Chemicals Supplier No. H3691103

CAS: 25028-32-4 | C10H11NO3 | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25028-32-4
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD11042531
InChI Key LUMZQFHOBXCXOR-UHFFFAOYSA-N
Synonym ethyl 2-acetylisonicotinate, 2-acetyl-4-pyridinecarboxylic acid ethyl ester, pyac32, 2-acetyl-isonicotinic acid ethyl ester, 2-acetylpyridine-4-carboxylic acid ethyl ester
PubChem CID 55265486
IUPAC Name ethyl 2-acetylpyridine-4-carboxylate
SMILES CCOC(=O)C1=CC(=NC=C1)C(C)=O

Specifications

Melting Point 52°C to 55°C
Color Cream
Quantity 1 g
Formula Weight 193.2
Percent Purity 96%
Physical Form Fused Solid
Chemical Name or Material Ethyl 2-acetylisonicotinate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only