CAS RN 25028-32-4
IUPAC Name:
ethyl 2-acetylpyridine-4-carboxylate
Synonyms:
ethyl 2-acetylpyridine-4-carboxylate
Molecular Weight (g/mol):
193.2
Molecular Formula:
C10H11NO3
InChi Key:
LUMZQFHOBXCXOR-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC(=NC=C1)C(C)=O
1
–
1
of
1
results
1
Promotions Available
Ethyl 2-acetylisonicotinate, 96%
Ethyl 2-acetylisonicotinate, 96%
CAS: 25028-32-4 Molecular Formula: C10H11NO3 Molecular Weight (g/mol): 193.20 MDL Number: MFCD11042531 InChI Key: LUMZQFHOBXCXOR-UHFFFAOYSA-N Synonym: ethyl 2-acetylisonicotinate,2-acetyl-4-pyridinecarboxylic acid ethyl ester,pyac32,2-acetyl-isonicotinic acid ethyl ester,2-acetylpyridine-4-carboxylic acid ethyl ester PubChem CID: 55265486 IUPAC Name: ethyl 2-acetylpyridine-4-carboxylate SMILES: CCOC(=O)C1=CC(=NC=C1)C(C)=O