missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Acetyl-3-ethoxyacrylate 98.0+%, TCI America™
$61.51 - $236.73
Chemical Identifiers
| CAS | 3788-94-1 |
|---|---|
| Molecular Formula | C9H14O4 |
| Molecular Weight (g/mol) | 186.207 |
| MDL Number | MFCD06797311 |
| InChI Key | FNASCUBBFNCFQO-VURMDHGXSA-N |
| Synonym | 2-Acetyl-3-ethoxyacrylic Acid Ethyl Ester |
| PubChem CID | 5355337 |
| IUPAC Name | ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate |
| SMILES | CCOC=C(C(=O)C)C(=O)OCC |
Chemical Identifiers
| 3788-94-1 | |
| 186.207 | |
| FNASCUBBFNCFQO-VURMDHGXSA-N | |
| 5355337 | |
| CCOC=C(C(=O)C)C(=O)OCC |
| C9H14O4 | |
| MFCD06797311 | |
| 2-Acetyl-3-ethoxyacrylic Acid Ethyl Ester | |
| ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate |
Specifications
| 3788-94-1 | |
| C9H14O4 | |
| 5 g | |
| FNASCUBBFNCFQO-VURMDHGXSA-N | |
| ethyl (2Z)-2-(ethoxymethylidene)-3-oxobutanoate | |
| 5355337 | |
| ≥98.0% (GC) | |
| Ethyl 2-Acetyl-3-ethoxyacrylate |
| 266°C | |
| MFCD06797311 | |
| 2-Acetyl-3-ethoxyacrylic Acid Ethyl Ester | |
| CCOC=C(C(=O)C)C(=O)OCC | |
| 186.207 | |
| 186.21 | |
| Liquid |
Safety and Handling
TSCA : No