missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-acetyl-3-(dimethylamino)acrylate, 95%, Thermo Scientific Chemicals
$880.89 - $880.89
Chemical Identifiers
| CAS | 51145-57-4 |
|---|---|
| Molecular Formula | C9H15NO3 |
| Molecular Weight (g/mol) | 185.223 |
| MDL Number | MFCD00087563 |
| InChI Key | LQSOVGAUOHMPLK-SOFGYWHQSA-N |
| PubChem CID | 2763877 |
| IUPAC Name | ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate |
| SMILES | CCOC(=O)C(=CN(C)C)C(=O)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6407314
|
Thermo Scientific Chemicals
H6407314 |
25 g |
Each for $880.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 51145-57-4 | |
| 185.223 | |
| LQSOVGAUOHMPLK-SOFGYWHQSA-N | |
| ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate |
| C9H15NO3 | |
| MFCD00087563 | |
| 2763877 | |
| CCOC(=O)C(=CN(C)C)C(=O)C |
Specifications
| 51145-57-4 | |
| C9H15NO3 | |
| 25 g | |
| LQSOVGAUOHMPLK-SOFGYWHQSA-N | |
| ethyl (2E)-2-(dimethylaminomethylidene)-3-oxobutanoate | |
| 2763877 | |
| 95% | |
| Ethyl 2-acetyl-3-(dimethylamino)acrylate |
| Red | |
| MFCD00087563 | |
| ethyl 2-acetyl-3-dimethylamino acrylate, e-ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl 2-dimethylamino methylene-3-oxobutanoate, ethyl n,n-dimethylaminomethylene acetoacetate, ethyl 2-dimethylamino methylidene-3-oxobutanoate, ethyl 2e-2-dimethylamino methylidene-3-oxobutanoate, 2-acetyl-3-dimethylamino-acrylic acid ethyl ester, ethyl 2e-2-acetyl-3-dimethylamino acrylate, ethyl 2e-2-dimethylaminomethylidene-3-oxobutanoate, ethyl2-acetyl-3-dimethylamino acrylate | |
| CCOC(=O)C(=CN(C)C)C(=O)C | |
| 185.223 | |
| 185.22 | |
| Liquid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only