missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoate 98.0+%, TCI America™

Catalog No. E06671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E06671G Supplier TCI America Supplier No. E06671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 346656-34-6
Molecular Formula C14H19BO4
Molecular Weight (g/mol) 262.112
InChI Key GZHASWCTAWNFFW-UHFFFAOYSA-N
Synonym 2-(2-Carbethoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane, 2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoic Acid Ethyl Ester, 2-(2-Ethoxycarbonylphenyl)-5,5-dimethyl-1,3,2-dioxaborinane
PubChem CID 10332902
IUPAC Name ethyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate
SMILES B1(OCC(CO1)(C)C)C2=CC=CC=C2C(=O)OCC

Specifications

Quantity 1 g
Formula Weight 262.11
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Ethyl 2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoate
TSCA No
Recommended Storage Refrigerator