accessibility menu, dialog, popup

UserName

CAS RN 346656-34-6

IUPAC Name: ethyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate
Synonyms: ethyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate
Molecular Weight (g/mol): 262.11
Molecular Formula: C14H19BO4
InChi Key: GZHASWCTAWNFFW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1B1OCC(C)(C)CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results