missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dioxo-5-methylhexanoate, 95%

Catalog No. p-7028421
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2573506 5 g
AAB2573514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2573506 Supplier Thermo Scientific Chemicals Supplier No. B2573506

CAS: 64195-85-3 | C9H14O4 | 186.21 g/mol

Ethyl 2,4-dioxo-5-methylhexanoate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2,4-dioxo-5-methylhexanoate is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 64195-85-3
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD07323638
InChI Key HBJPUWYQVWIKSD-UHFFFAOYSA-N
Synonym ethyl5-methyl-2,4-dioxohexanoate, ethyl 2,4-dioxo-5-methylhexanoate, 5-methyl-2,4-dioxohexanoic acid ethyl ester, hexanoic acid,5-methyl-2,4-dioxo-, ethyl ester, 5-methyl-2,4-dioxo-hexanoic acid ethyl ester, acmc-1bc7v, ethyl-2,4-dioxo-5-methylhexanoate, ethyl 5-methyl-2,4-dioxo-hexanoate, ethyl 5-methyl-2,4-bis oxidanylidene hexanoate
PubChem CID 3017501
IUPAC Name ethyl 5-methyl-2,4-dioxohexanoate
SMILES CCOC(=O)C(=O)CC(=O)C(C)C

Specifications

Quantity 5 g
Formula Weight 186.21
Percent Purity 95%
Chemical Name or Material Ethyl 2,4-dioxo-5-methylhexanoate
EINECSNumber 264-728-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only