accessibility menu, dialog, popup

UserName

H3COOOOCH3CH3

CAS RN 64195-85-3

IUPAC Name: ethyl 5-methyl-2,4-dioxohexanoate
Synonyms: ethyl 5-methyl-2,4-dioxohexanoate
Molecular Weight (g/mol): 186.21
Molecular Formula: C9H14O4
InChi Key: HBJPUWYQVWIKSD-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)CC(=O)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results